Product Details

SNP ID
rs149110871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4911615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACTTCGACCCCCTGAGGAACGA[A/G]TACTTCTTCGACCGCAACCGGCCCA
Phenotype
MIM: 176260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNA1 PubMed Links

Gene Details

Gene
KCNA1
Gene Name
potassium voltage-gated channel subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000217.2 1342 Silent Mutation GAA,GAG E79E NP_000208.2

View Full Product Details