Product Details

SNP ID
rs149119632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122238634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATCCAAAACTTTTTGTTTGAGCC[C/T]ACTATTACACACGGATTGGAGGCAA
Phenotype
MIM: 610034
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101593348 PubMed Links

Gene Details

Gene
LOC101593348
Gene Name
uncharacterized LOC101593348
There are no transcripts associated with this gene.

Gene
VPS33A
Gene Name
VPS33A, CORVET/HOPS core subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022916.4 1368 Missense Mutation AGG,GGG R419G NP_075067.2

View Full Product Details