Product Details

SNP ID
rs149323049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55321075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTAATCATATTCCCAGCACTCA[C/T]GTTGCTTTTAAAGCTTCATTACTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6C1 PubMed Links

Gene Details

Gene
OR6C1
Gene Name
olfactory receptor family 6 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005182.1 476 Missense Mutation ACG,ATG T159M NP_001005182.1

View Full Product Details