Product Details
- SNP ID
-
rs149748726
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:51189580 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCTGTTTTAGTGCAATTTTCCCC[A/G]CAAAGGAACCAGGAAGAGGTGGAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
POU6F1
PubMed Links
Gene Details
- Gene
- POU6F1
- Gene Name
- POU class 6 homeobox 1
View Full Product Details