Product Details

SNP ID
rs150010110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113468316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGTTGTTGGCCGTCTCCTTGTC[C/T]GACGAGGTCGAGTTGTCCGTCTCTT
Phenotype
MIM: 605992
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LHX5 PubMed Links

Gene Details

Gene
LHX5
Gene Name
LIM homeobox 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022363.2 1060 Silent Mutation TCA,TCG S162S NP_071758.1
Gene
LHX5-AS1
Gene Name
LHX5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details