Product Details

SNP ID
rs150615186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:82354512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTTACTTGTTTCTTAGCAGCAC[A/G]TTTGGCTTGTATCTGTTCATATTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC59 PubMed Links

Gene Details

Gene
CCDC59
Gene Name
coiled-coil domain containing 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014167.4 591 Missense Mutation CGT,TGT R183C NP_054886.2
Gene
METTL25
Gene Name
methyltransferase like 25
There are no transcripts associated with this gene.

View Full Product Details