Product Details

SNP ID
rs150864769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71440201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGCCCCACACACTGTCATTGC[A/G]AGCCCGACGGCAGGATGTTGCTCAG
Phenotype
MIM: 606667
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LGR5 PubMed Links

Gene Details

Gene
LGR5
Gene Name
leucine rich repeat containing G protein-coupled receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277226.1 432 Missense Mutation AAG,GAG K41E NP_001264155.1
NM_001277227.1 432 Missense Mutation AAG,GAG K41E NP_001264156.1
NM_003667.3 432 Missense Mutation AAG,GAG K41E NP_003658.1

View Full Product Details