Product Details

SNP ID
rs150921104
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68295333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGCCCGGAGACTAGAAGCTGCA[A/G]AGCGAGCAGAAATCTCAGACAATCT
Phenotype
MIM: 613813
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MDM1 PubMed Links

Gene Details

Gene
MDM1
Gene Name
Mdm1 nuclear protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205028.1 2094 Missense Mutation TCT,TTT S654F NP_001191957.1
NM_001205029.1 2094 Intron NP_001191958.1
NM_017440.4 2094 Missense Mutation TCT,TTT S689F NP_059136.2
NM_020128.2 2094 Intron NP_064513.1
XM_005269026.1 2094 Missense Mutation TCT,TTT S699F XP_005269083.1
XM_011538569.1 2094 Intron XP_011536871.1
XM_017019646.1 2094 Missense Mutation TCT,TTT S419F XP_016875135.1
XM_017019647.1 2094 Missense Mutation TCT,TTT S419F XP_016875136.1
XM_017019648.1 2094 Missense Mutation TCT,TTT S419F XP_016875137.1
XM_017019649.1 2094 Missense Mutation TCT,TTT S419F XP_016875138.1
XM_017019650.1 2094 Missense Mutation TCT,TTT S419F XP_016875139.1

View Full Product Details