Product Details

SNP ID
rs150994609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:101157358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAAAGCAGGATTATCAGTTCCA[C/T]CATCTTCCACTGGATGTGATTTATA
Phenotype
MIM: 608044
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC5A8 PubMed Links

Gene Details

Gene
SLC5A8
Gene Name
solute carrier family 5 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145913.3 2144 Missense Mutation GAT,GGT D585G NP_666018.3
XM_017018910.1 2144 Intron XP_016874399.1

View Full Product Details