Product Details

SNP ID
rs151008594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98734464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAATCAAAATTCAAAGATTTTTT[C/T]TGTTTCTGTAACTGGAACTAAATCA
Phenotype
MIM: 607815 MIM: 602233
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS1B PubMed Links

Gene Details

Gene
ANKS1B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204065.1 5153 Intron NP_001190994.1
NM_001204066.1 5153 Intron NP_001190995.1
NM_001204067.1 5153 Intron NP_001190996.1
NM_001204068.1 5153 Intron NP_001190997.1
NM_001204069.1 5153 Intron NP_001190998.1
NM_001204070.1 5153 Intron NP_001190999.1
NM_001204079.1 5153 Intron NP_001191008.1
NM_001204080.1 5153 Intron NP_001191009.1
NM_001204081.1 5153 Intron NP_001191010.1
NM_020140.3 5153 Intron NP_064525.1
NM_152788.4 5153 Intron NP_690001.3
NM_181670.3 5153 Intron NP_858056.2
XM_005269028.4 5153 Intron XP_005269085.1
XM_005269029.4 5153 Intron XP_005269086.1
XM_005269032.3 5153 Intron XP_005269089.1
XM_006719504.3 5153 Intron XP_006719567.1
XM_006719505.3 5153 Intron XP_006719568.1
XM_006719506.3 5153 Intron XP_006719569.1
XM_006719507.3 5153 UTR 3 XP_006719570.1
XM_006719508.3 5153 Intron XP_006719571.1
XM_006719509.3 5153 Intron XP_006719572.1
XM_006719510.3 5153 Intron XP_006719573.1
XM_006719512.3 5153 Intron XP_006719575.1
XM_006719513.3 5153 Intron XP_006719576.1
XM_006719514.3 5153 UTR 3 XP_006719577.1
XM_011538571.2 5153 Intron XP_011536873.1
XM_017019651.1 5153 Intron XP_016875140.1
XM_017019652.1 5153 Intron XP_016875141.1
XM_017019653.1 5153 UTR 3 XP_016875142.1
XM_017019654.1 5153 UTR 3 XP_016875143.1
XM_017019655.1 5153 Intron XP_016875144.1
XM_017019656.1 5153 Intron XP_016875145.1
XM_017019657.1 5153 Intron XP_016875146.1
XM_017019658.1 5153 UTR 3 XP_016875147.1
XM_017019659.1 5153 Intron XP_016875148.1
XM_017019660.1 5153 Intron XP_016875149.1
XM_017019661.1 5153 Intron XP_016875150.1
XM_017019662.1 5153 Intron XP_016875151.1
XM_017019663.1 5153 Intron XP_016875152.1
XM_017019664.1 5153 Intron XP_016875153.1
XM_017019665.1 5153 Intron XP_016875154.1
Gene
APAF1
Gene Name
apoptotic peptidase activating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160.2 5153 UTR 3 NP_001151.1
NM_013229.2 5153 UTR 3 NP_037361.1
NM_181861.1 5153 UTR 3 NP_863651.1
NM_181868.1 5153 UTR 3 NP_863658.1
NM_181869.1 5153 UTR 3 NP_863659.1
XM_017019250.1 5153 Intron XP_016874739.1

View Full Product Details