Product Details

SNP ID
rs151086994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53049150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGGAGGCCGGAGGCCCATGGAT[A/G]GGGGTGGAGTATGTGTTGGGAGGGG
Phenotype
MIM: 603928 MIM: 607717
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF4B PubMed Links

Gene Details

Gene
EIF4B
Gene Name
eukaryotic translation initiation factor 4B
There are no transcripts associated with this gene.

Gene
LOC283335
Gene Name
uncharacterized LOC283335
There are no transcripts associated with this gene.

Gene
MIR6757
Gene Name
microRNA 6757
There are no transcripts associated with this gene.

Gene
TNS2
Gene Name
tensin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015319.2 1561 Missense Mutation AGG,GGG R3G NP_056134.2
NM_170754.2 1561 Intron NP_736610.2
NM_198316.1 1561 Intron NP_938072.1
XM_006719302.3 1561 Missense Mutation AGG,GGG R3G XP_006719365.1
XM_006719303.1 1561 Intron XP_006719366.1
XM_011538079.1 1561 Intron XP_011536381.1
XM_017019088.1 1561 Intron XP_016874577.1
XM_017019089.1 1561 Intron XP_016874578.1

View Full Product Details