Product Details

SNP ID
rs151158281
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14882253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCACAGGCTTCCCTATTGAGCTC[A/G]TGGACAGGCTTAGAGCGTTCTCGGA
Phenotype
MIM: 154870
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MGP PubMed Links

Gene Details

Gene
MGP
Gene Name
matrix Gla protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000900.3 401 Silent Mutation CAC,CAT H66H NP_000891.2
NM_001190839.1 401 Silent Mutation CAC,CAT H91H NP_001177768.1

View Full Product Details