Product Details

SNP ID
rs140661222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110152438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACTCGATGAATGGCGCACTTCT[A/G]AACTCCTCCAGGCAGGAGCCGGGGG
Phenotype
MIM: 120130
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL4A1 PubMed Links

Gene Details

Gene
COL4A1
Gene Name
collagen type IV alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303110.1 12886 Intron NP_001290039.1
NM_001845.5 12886 Silent Mutation TTC,TTT F1608F NP_001836.3
XM_011521048.2 12886 Silent Mutation TTC,TTT F1544F XP_011519350.1

View Full Product Details