Product Details

SNP ID
rs141454852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:46342107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCACAGTTCTGGCAGAGGCACTG[C/G]GGATGAGAAAACAATTGACAGAATG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0226L PubMed Links

Gene Details

Gene
KIAA0226L
Gene Name
KIAA0226 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286761.1 3215 UTR 3 NP_001273690.1
NM_001286762.1 3215 UTR 3 NP_001273691.1
NM_001286763.1 3215 UTR 3 NP_001273692.1
NM_001286764.1 3215 UTR 3 NP_001273693.1
NM_001286765.1 3215 UTR 3 NP_001273694.1
NM_001286766.1 3215 UTR 3 NP_001273695.1
NM_025113.3 3215 UTR 3 NP_079389.2

View Full Product Details