Product Details

SNP ID
rs142530569
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:80336971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATCGTGTACAACAGTGAGACTG[A/G]CTGCAAGAACATGGGTTGTCAGCAC
Phenotype
MIM: 602466
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SPRY2 PubMed Links

Gene Details

Gene
SPRY2
Gene Name
sprouty RTK signaling antagonist 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318536.1 1280 Silent Mutation AGC,AGT S245S NP_001305465.1
NM_001318537.1 1280 Silent Mutation AGC,AGT S245S NP_001305466.1
NM_001318538.1 1280 Silent Mutation AGC,AGT S245S NP_001305467.1
NM_005842.3 1280 Silent Mutation AGC,AGT S245S NP_005833.1

View Full Product Details