Product Details

SNP ID
rs146726513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102769073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTCCAGGGAGAAAAATCATAAG[C/T]AGCTTCCTTGGCAATATCACGGAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEX30 PubMed Links

Gene Details

Gene
TEX30
Gene Name
testis expressed 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286775.1 133 Intron NP_001273704.1
NM_001286776.1 133 Intron NP_001273705.1
NM_138779.4 133 Intron NP_620134.3
XM_005254097.2 133 Intron XP_005254154.1
XM_011521135.2 133 Intron XP_011519437.1
XM_011521136.2 133 Missense Mutation ACT,GCT T11A XP_011519438.1
XM_017020852.1 133 Intron XP_016876341.1

View Full Product Details