Product Details

SNP ID
rs147666396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:78615997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGAGAACCTTTGGAAAGGTTTAG[C/T]TCAGTTTTTTCTTCCAACTTATCAG
Phenotype
MIM: 615906
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF219 PubMed Links

Gene Details

Gene
RNF219
Gene Name
ring finger protein 219
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024546.3 1823 Silent Mutation GAA,GAG E588E NP_078822.3
XM_011535225.1 1823 Silent Mutation GAA,GAG E511E XP_011533527.1
XM_017020743.1 1823 Silent Mutation GAA,GAG E511E XP_016876232.1
Gene
RNF219-AS1
Gene Name
RNF219 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details