Product Details

SNP ID
rs148206819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52378140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGGCACAACTGGCCTGAGTCAC[A/G]TTTAGATCCAGCCTGGAGGGAGGCC
Phenotype
MIM: 616821
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
THSD1 PubMed Links

Gene Details

Gene
THSD1
Gene Name
thrombospondin type 1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018676.3 2375 Silent Mutation AAC,AAT N610N NP_061146.1
NM_199263.2 2375 Silent Mutation AAC,AAT N557N NP_954872.1

View Full Product Details