Product Details
- SNP ID
-
rs148307139
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:102687013 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGTATTTTTGCATCTTGGAAATT[C/G]AATTCCTCGGCATGTTCCTCCAAGT
- Phenotype
-
MIM: 615259
MIM: 190470
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
METTL21C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs660207] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- METTL21C
- Gene Name
- methyltransferase like 21C
- Gene
- TPP2
- Gene Name
- tripeptidyl peptidase 2
There are no transcripts associated with this gene.
View Full Product Details