Product Details

SNP ID
rs149407344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110308518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGAACTCGGGAGCTGGTGGGGA[A/G]TCTCTGTCACGGACTGAGGGTTTTT
Phenotype
MIM: 120130 MIM: 120090
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL4A1 PubMed Links

Gene Details

Gene
COL4A1
Gene Name
collagen type IV alpha 1 chain
There are no transcripts associated with this gene.

Gene
COL4A2
Gene Name
collagen type IV alpha 2
There are no transcripts associated with this gene.

View Full Product Details