Product Details

SNP ID
rs149654122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49507597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAATTGTTCTTCAAAGAGATTGT[A/G]CCATTTAATACACCTAACTTACCAA
Phenotype
MIM: 607796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHF11 PubMed Links

Gene Details

Gene
PHF11
Gene Name
PHD finger protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040443.2 Intron NP_001035533.1
NM_001040444.2 Intron NP_001035534.1
Gene
SETDB2-PHF11
Gene Name
SETDB2-PHF11 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320727.1 Intron NP_001307656.1

View Full Product Details