Product Details

SNP ID
rs150083496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:114242152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCTATCTGTCTTCTACGCGGGA[A/G]AATCTATGATGCTCTAGATAACCGA
Phenotype
MIM: 603461
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC16 PubMed Links

Gene Details

Gene
CDC16
Gene Name
cell division cycle 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005266206.1 598 Missense Mutation AAA,AGA K138R XP_005266263.1
XM_006719992.1 598 Missense Mutation AAA,AGA K137R XP_006720055.1
XM_006719993.1 598 Missense Mutation AAA,AGA K44R XP_006720056.1
XM_011534851.1 598 Missense Mutation AAA,AGA K138R XP_011533153.1
XM_011534853.1 598 Missense Mutation AAA,AGA K138R XP_011533155.1
XM_017020830.1 598 Missense Mutation AAA,AGA K138R XP_016876319.1
XM_017020831.1 598 Missense Mutation AAA,AGA K138R XP_016876320.1
XM_017020832.1 598 Missense Mutation AAA,AGA K44R XP_016876321.1
XM_017020833.1 598 Missense Mutation AAA,AGA K44R XP_016876322.1
Gene
MIR548AR
Gene Name
microRNA 548ar
There are no transcripts associated with this gene.

View Full Product Details