Product Details

SNP ID
rs150173381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:46345536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTCTTGATCCTGACCAGGTCCT[C/T]AAGGGAGAACAGGTGGAGCTCATCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0226L PubMed Links

Gene Details

Gene
KIAA0226L
Gene Name
KIAA0226 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286761.1 1644 Missense Mutation AAG,GAG K566E NP_001273690.1
NM_001286762.1 1644 Intron NP_001273691.1
NM_001286763.1 1644 Missense Mutation AAG,GAG K499E NP_001273692.1
NM_001286764.1 1644 Missense Mutation AAG,GAG K431E NP_001273693.1
NM_001286765.1 1644 Missense Mutation AAG,GAG K409E NP_001273694.1
NM_001286766.1 1644 Missense Mutation AAG,GAG K351E NP_001273695.1
NM_025113.3 1644 Missense Mutation AAG,GAG K566E NP_079389.2

View Full Product Details