Product Details

SNP ID
rs138179143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21155011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGTTTCTCAAAGAGTACATGA[A/G]GGGGTTCAGCACTGGGATCACCACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5AU1 PubMed Links

Gene Details

Gene
OR5AU1
Gene Name
olfactory receptor family 5 subfamily AU member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004731.1 1015 Missense Mutation ATC,GTC I339V NP_001004731.1

View Full Product Details