Product Details

SNP ID
rs139780666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:95183905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCTCCCACGATTGTTCTAAGGG[C/T]CAAGTGAGTTAATGAGTAAACAATG
Phenotype
MIM: 611121
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLMN PubMed Links

Gene Details

Gene
CLMN
Gene Name
calmin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024734.3 11043 UTR 3 NP_079010.2
XM_011537158.1 11043 UTR 3 XP_011535460.1
XM_011537159.2 11043 Intron XP_011535461.1
XM_017021646.1 11043 UTR 3 XP_016877135.1
XM_017021647.1 11043 Intron XP_016877136.1
Gene
DICER1-AS1
Gene Name
DICER1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details