Product Details

SNP ID
rs140603938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21468435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTCTAACCATGAGGTCAGGTGG[C/T]TGAAGGTTTCACGCCTACAGCAGAA
Phenotype
MIM: 607466 MIM: 614032
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB2B PubMed Links

Gene Details

Gene
RAB2B
Gene Name
RAB2B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163380.1 312 Missense Mutation AAC,AGC N49S NP_001156852.1
NM_032846.3 312 Missense Mutation AAC,AGC N95S NP_116235.2
XM_017021710.1 312 Missense Mutation AAC,AGC N50S XP_016877199.1
XM_017021711.1 312 Missense Mutation AAC,AGC N30S XP_016877200.1
XM_017021712.1 312 Missense Mutation AAC,AGC N26S XP_016877201.1
Gene
TOX4
Gene Name
TOX high mobility group box family member 4
There are no transcripts associated with this gene.

View Full Product Details