Product Details

SNP ID
rs140687294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24299729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCGATTCTGTGCTGAGGTAGAG[A/G]GGCAGAGTCCCAGGCCAAAGTTAGA
Phenotype
MIM: 604441 MIM: 610410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIDEB PubMed Links

Gene Details

Gene
CIDEB
Gene Name
cell death-inducing DFFA-like effector b
There are no transcripts associated with this gene.

Gene
DHRS1
Gene Name
dehydrogenase/reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136050.2 105 UTR 5 NP_001129522.1
NM_138452.2 105 Intron NP_612461.1
Gene
NOP9
Gene Name
NOP9 nucleolar protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286367.1 105 Intron NP_001273296.1
NM_174913.2 105 Intron NP_777573.1
XM_005267385.1 105 Intron XP_005267442.1
XM_011536526.1 105 Intron XP_011534828.1
XM_011536527.2 105 Intron XP_011534829.1

View Full Product Details