Product Details

SNP ID
rs140798864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103519897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGGCATGAGGTCGTCGATGGC[C/T]TGGCCCTGCTCCAGCCGCTGCTCCA
Phenotype
MIM: 123280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CKB PubMed Links

Gene Details

Gene
CKB
Gene Name
creatine kinase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001823.4 1265 Silent Mutation CAA,CAG Q371Q NP_001814.2
XM_017020951.1 1265 Silent Mutation CAA,CAG Q395Q XP_016876440.1
XM_017020952.1 1265 Intron XP_016876441.1
Gene
TRMT61A
Gene Name
tRNA methyltransferase 61A
There are no transcripts associated with this gene.

View Full Product Details