Product Details

SNP ID
rs141089655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60734923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATATCGGAACCCCTCCTTGAAG[C/G]TGATGGTGAATGTGTGCGGACACAC
Phenotype
MIM: 602659
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MNAT1 PubMed Links

Gene Details

Gene
MNAT1
Gene Name
MNAT1, CDK activating kinase assembly factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177963.1 189 Silent Mutation CTG,GTG L21V NP_001171434.1
NM_002431.3 189 Silent Mutation CTG,GTG L21V NP_002422.1
XM_005267687.2 189 Missense Mutation CTG,GTG L21V XP_005267744.1
XM_005267688.2 189 Missense Mutation CTG,GTG L21V XP_005267745.1
XM_017021332.1 189 Intron XP_016876821.1
XM_017021333.1 189 UTR 5 XP_016876822.1
XM_017021334.1 189 Intron XP_016876823.1

View Full Product Details