Product Details

SNP ID
rs141350994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:31483304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTACTCTTTCATAACAAAAGATG[A/T]TGATGAAGAAAGGCAGAAGAAAGCC
Phenotype
MIM: 610118
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR33 PubMed Links

Gene Details

Gene
GPR33
Gene Name
G protein-coupled receptor 33 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197184.2 717 Missense Mutation AAC,ATC N221I NP_001184113.2

View Full Product Details