Product Details

SNP ID
rs141396281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100381294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTGTGGACGAGCCATGTTCCA[A/G]CCAGCCACATGCCCCTGGCAGCTGC
Phenotype
MIM: 191050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WARS PubMed Links

Gene Details

Gene
WARS
Gene Name
tryptophanyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
WDR25
Gene Name
WD repeat domain 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161476.1 470 Missense Mutation ACC,GCC T124A NP_001154948.1
NM_024515.4 470 Missense Mutation ACC,GCC T124A NP_078791.3
XM_005268056.4 470 Missense Mutation ACC,GCC T124A XP_005268113.1
XM_005268057.3 470 Intron XP_005268114.1
XM_005268058.4 470 Intron XP_005268115.1
XM_011537150.2 470 Missense Mutation ACC,GCC T124A XP_011535452.1
XM_011537151.2 470 Intron XP_011535453.2
XM_017021639.1 470 Intron XP_016877128.1

View Full Product Details