Product Details

SNP ID
rs142908236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:90873671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACACAGGTATGCACGGCAGCTCC[A/G]GAAGATCCTAGAATATCCGGAGTCA
Phenotype
MIM: 603607
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPS6KA5 PubMed Links

Gene Details

Gene
RPS6KA5
Gene Name
ribosomal protein S6 kinase A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322227.1 2228 Silent Mutation NP_001309156.1
NM_001322228.1 2228 Silent Mutation NP_001309157.1
NM_001322229.1 2228 Silent Mutation NP_001309158.1
NM_001322230.1 2228 Silent Mutation NP_001309159.1
NM_001322231.1 2228 Silent Mutation NP_001309160.1
NM_001322232.1 2228 Silent Mutation NP_001309161.1
NM_001322233.1 2228 Silent Mutation NP_001309162.1
NM_001322234.1 2228 Silent Mutation NP_001309163.1
NM_001322235.1 2228 Silent Mutation NP_001309164.1
NM_001322236.1 2228 Silent Mutation NP_001309165.1
NM_001322237.1 2228 Silent Mutation NP_001309166.1
NM_001322238.1 2228 Silent Mutation NP_001309167.1
NM_004755.3 2228 Silent Mutation NP_004746.2
NM_182398.2 2228 Intron NP_872198.1
XM_017021786.1 2228 Silent Mutation XP_016877275.1

View Full Product Details