Product Details

SNP ID
rs143017999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:98716307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCAGTGAGCCAACTTGGTGAAG[A/G]GAAATGCCCAAGTTCTCAGCACCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf177 PubMed Links

Gene Details

Gene
C14orf177
Gene Name
chromosome 14 open reading frame 177
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182560.2 535 Missense Mutation GAG,GGG E39G NP_872366.2

View Full Product Details