Product Details

SNP ID
rs143234123
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23059212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCGGCTTCTGCTGTGGCGCTTG[A/G]TGTCCCTGCGATCCCTTTCCCTGCC
Phenotype
MIM: 604562
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ACIN1 PubMed Links

Gene Details

Gene
ACIN1
Gene Name
apoptotic chromatin condensation inducer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164814.1 4091 Missense Mutation NP_001158286.1
NM_001164815.1 4091 Missense Mutation NP_001158287.1
NM_001164816.1 4091 Missense Mutation NP_001158288.1
NM_001164817.1 4091 Missense Mutation NP_001158289.1
NM_014977.3 4091 Missense Mutation NP_055792.1
XM_005267415.3 4091 Missense Mutation XP_005267472.1
XM_005267416.3 4091 Missense Mutation XP_005267473.1
XM_005267418.1 4091 Missense Mutation XP_005267475.1
XM_006720081.3 4091 Intron XP_006720144.1
XM_011536570.2 4091 Intron XP_011534872.1
Gene
CDH24
Gene Name
cadherin 24
There are no transcripts associated with this gene.

Gene
LOC105370705
Gene Name
collagen alpha-2(I) chain-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011537450.1 4091 Intron XP_011535752.1

View Full Product Details