Product Details
- SNP ID
-
rs143379381
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:22987495 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTGTGAGGCGCTGGGCCCGGAAA[C/T]GGGGAGGGCGGGCCACCCACACACC
- Phenotype
-
MIM: 609066
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AJUBA
PubMed Links
Gene Details
- Gene
- AJUBA
- Gene Name
- ajuba LIM protein
There are no transcripts associated with this gene.
- Gene
- C14orf93
- Gene Name
- chromosome 14 open reading frame 93
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130706.2 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
NP_001124178.1 |
NM_001130708.2 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
NP_001124180.1 |
NM_001282968.1 |
1516 |
Missense Mutation |
CAT,CGT |
H266R |
NP_001269897.1 |
NM_001282969.1 |
1516 |
Missense Mutation |
CAT,CGT |
H266R |
NP_001269898.1 |
NM_001282970.1 |
1516 |
Intron |
|
|
NP_001269899.1 |
NM_021944.3 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
NP_068763.2 |
XM_005267971.1 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
XP_005268028.1 |
XM_005267972.1 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
XP_005268029.1 |
XM_006720231.1 |
1516 |
Missense Mutation |
CAT,CGT |
H266R |
XP_006720294.1 |
XM_006720232.2 |
1516 |
Missense Mutation |
CAT,CGT |
H266R |
XP_006720295.1 |
XM_011537059.1 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
XP_011535361.1 |
XM_011537060.2 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
XP_011535362.1 |
XM_011537062.1 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
XP_011535364.1 |
XM_017021569.1 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
XP_016877058.1 |
XM_017021570.1 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
XP_016877059.1 |
XM_017021571.1 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
XP_016877060.1 |
XM_017021572.1 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
XP_016877061.1 |
XM_017021573.1 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
XP_016877062.1 |
XM_017021574.1 |
1516 |
Missense Mutation |
CAT,CGT |
H446R |
XP_016877063.1 |
XM_017021575.1 |
1516 |
Missense Mutation |
CAT,CGT |
H266R |
XP_016877064.1 |
XM_017021576.1 |
1516 |
Missense Mutation |
CAT,CGT |
H266R |
XP_016877065.1 |
XM_017021577.1 |
1516 |
Missense Mutation |
CAT,CGT |
H266R |
XP_016877066.1 |
View Full Product Details