Product Details

SNP ID
rs143432605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:101562043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCGCATGAGCGCCTTCCAGCGC[C/G]TGGTCACTAAGTACCAGCGCGACGT
Phenotype
MIM: 601038 MIM: 608523
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DIO3 PubMed Links

Gene Details

Gene
DIO3
Gene Name
deiodinase, iodothyronine, type III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001362.3 693 Missense Mutation CTG,GTG L183V NP_001353.4
Gene
DIO3OS
Gene Name
DIO3 opposite strand/antisense RNA (head to head)
There are no transcripts associated with this gene.

Gene
MIR1247
Gene Name
microRNA 1247
There are no transcripts associated with this gene.

View Full Product Details