Product Details

SNP ID
rs143703299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60436450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAATTTTTAGTAGCTAAAGTAC[A/G]CTTCAGCACTAACTCTGGAGTTACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf39 PubMed Links

Gene Details

Gene
C14orf39
Gene Name
chromosome 14 open reading frame 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174978.2 2232 UTR 3 NP_777638.2
XM_011536690.2 2232 UTR 3 XP_011534992.1
XM_011536703.2 2232 Intron XP_011535005.1
XM_011536704.1 2232 Intron XP_011535006.1
XM_017021247.1 2232 UTR 3 XP_016876736.1
XM_017021248.1 2232 UTR 3 XP_016876737.1
XM_017021249.1 2232 UTR 3 XP_016876738.1
XM_017021250.1 2232 UTR 3 XP_016876739.1
XM_017021251.1 2232 UTR 3 XP_016876740.1

View Full Product Details