Product Details

SNP ID
rs143776833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24319201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATAGTGCCAAGGTGGCTGCAGCT[A/C]CGTTCAGCATCCTGGCAATGGGCCC
Phenotype
MIM: 600292 MIM: 604441 MIM: 601531 MIM: 605773
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADCY4 PubMed Links

Gene Details

Gene
ADCY4
Gene Name
adenylate cyclase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198568.1 2971 Silent Mutation CGG,CGT R951R NP_001185497.1
NM_001198592.1 2971 Silent Mutation CGG,CGT R951R NP_001185521.1
NM_139247.3 2971 Silent Mutation CGG,CGT R951R NP_640340.2
Gene
CIDEB
Gene Name
cell death-inducing DFFA-like effector b
There are no transcripts associated with this gene.

Gene
LTB4R
Gene Name
leukotriene B4 receptor
There are no transcripts associated with this gene.

Gene
LTB4R2
Gene Name
leukotriene B4 receptor 2
There are no transcripts associated with this gene.

View Full Product Details