Product Details
- SNP ID
-
rs143846565
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:90872089 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTACCATGCCTAAGCTACTGAGTC[C/T]GAGAACTGGAAGAGGGTCTCCGGGT
- Phenotype
-
MIM: 603607
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RPS6KA5
PubMed Links
Gene Details
- Gene
- RPS6KA5
- Gene Name
- ribosomal protein S6 kinase A5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001322227.1 |
2501 |
Silent Mutation |
TCA,TCG |
S517S |
NP_001309156.1 |
NM_001322228.1 |
2501 |
Silent Mutation |
TCA,TCG |
S762S |
NP_001309157.1 |
NM_001322229.1 |
2501 |
Silent Mutation |
TCA,TCG |
S791S |
NP_001309158.1 |
NM_001322230.1 |
2501 |
Silent Mutation |
TCA,TCG |
S531S |
NP_001309159.1 |
NM_001322231.1 |
2501 |
Silent Mutation |
TCA,TCG |
S559S |
NP_001309160.1 |
NM_001322232.1 |
2501 |
Silent Mutation |
TCA,TCG |
S725S |
NP_001309161.1 |
NM_001322233.1 |
2501 |
Silent Mutation |
TCA,TCG |
S744S |
NP_001309162.1 |
NM_001322234.1 |
2501 |
Silent Mutation |
TCA,TCG |
S606S |
NP_001309163.1 |
NM_001322235.1 |
2501 |
Silent Mutation |
TCA,TCG |
S719S |
NP_001309164.1 |
NM_001322236.1 |
2501 |
Silent Mutation |
TCA,TCG |
S770S |
NP_001309165.1 |
NM_001322237.1 |
2501 |
Silent Mutation |
TCA,TCG |
S719S |
NP_001309166.1 |
NM_001322238.1 |
2501 |
Silent Mutation |
TCA,TCG |
S579S |
NP_001309167.1 |
NM_004755.3 |
2501 |
Silent Mutation |
TCA,TCG |
S798S |
NP_004746.2 |
NM_182398.2 |
2501 |
Intron |
|
|
NP_872198.1 |
XM_017021786.1 |
2501 |
Silent Mutation |
TCA,TCG |
S579S |
XP_016877275.1 |
View Full Product Details