Product Details

SNP ID
rs144529882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20956202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGCATGTGACAACAGAGATCAAC[C/G]ACGAGACCCTCCACAGTATCCGGTG
Phenotype
MIM: 131410
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNASE2 PubMed Links

Gene Details

Gene
RNASE2
Gene Name
ribonuclease A family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002934.2 502 Missense Mutation CCA,CGA P144R NP_002925.1

View Full Product Details