Product Details

SNP ID
rs145124379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77474947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTCCCTGCCTGGAGTCCACTTC[A/G]CCTCAGCTGGGCTGGCTTGCAGCCC
Phenotype
MIM: 608466 MIM: 612684
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AHSA1 PubMed Links

Gene Details

Gene
AHSA1
Gene Name
activator of Hsp90 ATPase activity 1
There are no transcripts associated with this gene.

Gene
ISM2
Gene Name
isthmin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182509.3 2328 UTR 3 NP_872315.2
NM_199296.2 2328 UTR 3 NP_954993.1
XM_011536489.1 2328 UTR 3 XP_011534791.1

View Full Product Details