Product Details

SNP ID
rs145245060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21461280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGATGTTGAAATTGACTGTTG[A/G]GGCCCAATCTTGATGCCATTTGCCT
Phenotype
MIM: 607466
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB2B PubMed Links

Gene Details

Gene
RAB2B
Gene Name
RAB2B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163380.1 595 Silent Mutation CCC,CCT P143P NP_001156852.1
NM_032846.3 595 Silent Mutation CCC,CCT P189P NP_116235.2
XM_017021710.1 595 Silent Mutation CCC,CCT P144P XP_016877199.1
XM_017021711.1 595 Silent Mutation CCC,CCT P124P XP_016877200.1
XM_017021712.1 595 Silent Mutation CCC,CCT P120P XP_016877201.1

View Full Product Details