Product Details

SNP ID
rs145265355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23475177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTTTTGTTCAACTCAGGGTCTC[A/G]GCTTCTTGGAAGTGAAAGACCAGCT
Phenotype
MIM: 610777
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NGDN PubMed Links

Gene Details

Gene
NGDN
Gene Name
neuroguidin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042635.1 178 Missense Mutation AGC,GGC S51G NP_001036100.1
NM_015514.1 178 Missense Mutation AGC,GGC S51G NP_056329.1

View Full Product Details