Product Details

SNP ID
rs145620507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:98716272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATAAGCAGGGAACAAAGCCGAT[C/G]ATCACACGCCCATCAGTGAGCCAAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C14orf177 PubMed Links

Gene Details

Gene
C14orf177
Gene Name
chromosome 14 open reading frame 177
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182560.2 500 Missense Mutation ATC,ATG I27M NP_872366.2

View Full Product Details