Product Details

SNP ID
rs145953540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60648641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACGCCGCGTACCTTTCCTTGGC[C/T]TCCGCGGCCCGGTCTCTTTGCCTCC
Phenotype
MIM: 601205
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105378189 PubMed Links

Gene Details

Gene
LOC105378189
Gene Name
uncharacterized LOC105378189
There are no transcripts associated with this gene.

Gene
SALRNA1
Gene Name
senescence associated long non-coding RNA 1
There are no transcripts associated with this gene.

Gene
SIX1
Gene Name
SIX homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005982.3 797 Silent Mutation GAA,GAG E183E NP_005973.1
XM_017021602.1 797 Intron XP_016877091.1

View Full Product Details