Product Details

SNP ID
rs145962477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50533112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCTGCACTGCACACCTGCACAA[C/T]CTGCCTTCTACTTAGTTCTTCTGAG
Phenotype
MIM: 606439 MIM: 604923
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATL1 PubMed Links

Gene Details

Gene
ATL1
Gene Name
atlastin GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127713.1 31 UTR 5 NP_001121185.1
NM_015915.4 31 Intron NP_056999.2
NM_181598.3 31 Intron NP_853629.2
Gene
MAP4K5
Gene Name
mitogen-activated protein kinase kinase kinase kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006575.4 31 Intron NP_006566.2
NM_198794.2 31 Intron NP_942089.1
XM_006720013.3 31 Intron XP_006720076.1
XM_011536377.2 31 Intron XP_011534679.1
XM_011536378.2 31 Intron XP_011534680.1
XM_011536379.2 31 Intron XP_011534681.1
XM_011536380.2 31 Intron XP_011534682.1

View Full Product Details