Product Details

SNP ID
rs145994294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52730529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCACAGTGCAAGGAAGACGCC[A/G]AGGTGAGTCGCTCCCGTGGCTGCCA
Phenotype
MIM: 602708 MIM: 615814
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMC6 PubMed Links

Gene Details

Gene
PSMC6
Gene Name
proteasome 26S subunit, ATPase 6
There are no transcripts associated with this gene.

Gene
STYX
Gene Name
serine/threonine/tyrosine interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130701.1 365 Missense Mutation AAG,GAG K19E NP_001124173.1
NM_145251.3 365 Missense Mutation AAG,GAG K19E NP_660294.1
XM_006720242.3 365 Intron XP_006720305.1
XM_011537108.2 365 Intron XP_011535410.1
XM_011537109.2 365 Intron XP_011535411.1

View Full Product Details