Product Details

SNP ID
rs146193175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:72974171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAATGAGCGTTCCACCTTCATC[A/G]TCCACTTGTGCCTCGGTAACAGCTG
Phenotype
MIM: 605471
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFYVE1 PubMed Links

Gene Details

Gene
ZFYVE1
Gene Name
zinc finger FYVE-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281734.1 867 Silent Mutation GAC,GAT D656D NP_001268663.1
NM_001281735.1 867 Silent Mutation GAC,GAT D255D NP_001268664.1
NM_021260.3 867 Silent Mutation GAC,GAT D670D NP_067083.1
NM_178441.2 867 Silent Mutation GAC,GAT D255D NP_848535.1
XM_017021373.1 867 Silent Mutation GAC,GAT D255D XP_016876862.1

View Full Product Details