Product Details

SNP ID
rs146202153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93183958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAAATGTATTATCTGGGTCAGG[A/G]GGCTTAAAGATCACTCTCCAGATAC
Phenotype
MIM: 609485
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOAP1 PubMed Links

Gene Details

Gene
MOAP1
Gene Name
modulator of apoptosis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022151.4 655 Silent Mutation CCC,CCT P95P NP_071434.2
Gene
TMEM251
Gene Name
transmembrane protein 251
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098621.1 655 Intron NP_001092091.1
NM_015676.1 655 Intron NP_056491.1

View Full Product Details