Product Details

SNP ID
rs147195510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24182002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTGCCAGCACGCCCATCCCGAA[A/G]ATGGCATTGCTTCGCACCTCGGGGT
Phenotype
MIM: 608193
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IPO4 PubMed Links

Gene Details

Gene
IPO4
Gene Name
importin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024658.3 2891 Silent Mutation ATC,ATT I920I NP_078934.3
Gene
REC8
Gene Name
REC8 meiotic recombination protein
There are no transcripts associated with this gene.

Gene
TM9SF1
Gene Name
transmembrane 9 superfamily member 1
There are no transcripts associated with this gene.

View Full Product Details